Learn More
Micronova™ MicroSeal™ Tape
For plate sealing, bundling and labeling applications
Brand:  Micronova™ CSLB-12WH
 View more versions of this product
Product Code. 13177316
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For plate sealing, bundling and labeling applications
  • Excellent peel/reseal properties
  • Easy tear perforations
  • Acrylic adhesive won't transfer
  • Can be written on with fiber tip and permanent markers
  • All rolls are 18 yd. (16.5m) long
Specifications
Specifications
White
Polyethylene
Non-sterile
1 Ea.
16.5 m
Tape
MicroSeal™ Tape
Aseptic gowning, Hazmat and safety gowning, Wheel taping and equipment binding
18 yd.
0.5 in.