Learn More
Thermo Scientific™ Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Integrate these microplate carriers with the microplate swinging bucket rotor.
Brand: Thermo Scientific™ 75003625
Code : 50
Additional Details : Weight : 1.13580kg
Description
Integrate Thermo Scientific™ Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor to add flexibility to the swinging-bucket rotor.
Includes:
Sealed carriers include trays and lids (set of 2). Replacement O-rings for lids are offered as a set of four, with grease.
Specifications
Microplate Carrier | |
Sealed Microplate Carriers | |
Biocontainment certified by CAMR™, HPA, Porton Down, U.K. |