missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Microplate Immuno Polysorp 96 Well
Brand: Thermo Scientific™ Nunc™ 473245
786.70 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Transferable solid phase (TSP) system facilitates the testing by an ELISA method of two types of analytes simultaneouslyUniform adsorption properties
Ideal for screening of hybridoma cultures for antibody production.
Specifications
Standard | |
Clear | |
96 | |
Round Bottom (U-well) |
8 x 12 | |
Polystyrene | |
PolySorp |