Learn More
Thermo Scientific™ Microplate Reader Verification Plates
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
£2670.00 - £3825.00
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
11958995
|
Thermo Scientific™
24072800 |
Multiskan Verification Plate, 8 wavelengths | NPL (National Physical Laboratory) traceable Multiskan™ Verification Plate Certificate of Calibration USB memory device including SkanIt™ software sessions and reference files Carrying case for Multiskan™ Verification Plate |
£2670.00
Each |
Estimated Shipment: 27-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12463458
|
Thermo Scientific™
2806460 |
Verification Plate | Lumiwell Luminometric Verification Plate Certificate of Calibration Carrying case Lumiwell Verification Plate |
£3825.00
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11560715
|
Thermo Scientific™
N03394 |
Spectrophotometric Verification Plate | NPL (UK National Physical Laboratory) and NIST (US National Institute for Standards and Technology) traceable Spectrophotometric Verification Plate Certificate of Calibration Adapter for Spectrophotometric Verification Plate USB memory device including SkanIt™ software sessions and reference files Carrying case for Spectrophotometric Verification Plate |
£2790.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17104669
|
Thermo Scientific™
24072805 |
Multiskan Verification Plate, 16 wavelengths | NPL (National Physical Laboratory) traceable Multiskan™ Verification Plate Certificate of Calibration USB memory device including SkanIt™ software sessions and reference files Carrying case for Multiskan™ Verification Plate |
£3225.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Multiskan Verification Plates and Spectrophotometric Verification Plate enables you to verify parameters such as photometric accuracy, precision and linearity, as well as wavelength accuracy and stray light radiation.Lumiwell Luminometric Verification Plate is designed to be an independent Relative Light Unit reference for luminometry with the Thermo Scientific luminescence microplate readers.
We recommend recalibrating the verification plates regularly. The frequency of calibration is dependent on use and laboratory procedures. However, the recalibration should be performed at least every two years.
Specifications
Verification Plate |
For Research Use Only. Not for use in diagnostic procedures.