missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Preparation Rack, 96-well, with cover

Thermo Scientific™ 96-well sample preparation rack.

Brand:  Thermo Scientific™ PRK0963

Product Code. 10475779

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ 96-well sample preparation rack.

For all 0.2mL tubes, strip tubes, 24-well Piko™ PCR Plates and 96-well non-skirted or semi-skirted plates.

  • Blue rack, clear cover
  • Industry-standard footprint fits in microplate centrifuge rotors
  • Clear lid allows for easy stacking and viewing of samples

Includes:

Recommended for:
Sample handling, sample preparation, sample storage

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Microplate Rack
0.2 mL Tubes
Real-Time PCR Systems or Thermal Cyclers
5 racks
Blue/Clear, Clear
Rectangular
5 Racks
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Preparation Rack, 96-well, with cover > Sample Prep Rack with lid; 5/Cs.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.