Learn More
Invitrogen™ Microplates for Fluorescence-based Assays
Microplates for Fluorescence-based Assays are black-walled, clear-bottom 384-well microplates that minimize well-to-well crosstalk and autofluorescence. These microplates are optimized for fluorescence-based applications including nucleic acid quantitation (e.g. Quant-iT assays).
£161.00
Product Code | Brand | No. of Wells | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Quantity | Price | Quantity & Availability | ||||
15329413
|
Invitrogen™
M33089 |
96 | 10 plates |
£161.00
Pack of 10 |
In Stock
Log in to see stock. |
||||
Description
Molecular Probes® 96-well microplates for fluorescence-based assays are black-walled, clear-bottom 96-well microplates that minimize well-to-well crosstalk and autofluorescence and so are optimized for fluorescence-based applications. These plates are compatible with standard fluorescence microplate readers and are offered in a convenient 10-pack size for those times when only a few assay plates are required.
- Black-walled, clear-bottom 96-well microplates that minimize well-to-well crosstalk and autofluorescence
- Compatible with standard fluorescence microplate readers
- Packaged in a convenient 10-pack size
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function
Specifications
Nunc | |
Non-sterile |
For Research Use Only. Not for use in diagnostic procedures.