missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ MicroPlus Membranes

Features very narrow pore size distribution, low levels of extractables and increased temperature stability. Whatman™ MicroPlus Membranes are suitable for sample preparation, microbiological studies and filtration of aqueous solutions.

Brand:  Cytiva 10407713

 View more versions of this product

Product Code. 11909217

  • £155.00 / Pack of 100
Estimated Shipment: 06-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The membrane filtration (MF) method is excellent for the determination and enumeration of microbial contaminants.
  • Special membrane filters made from cellulose nitrate developed for microbiological quality control
  • High mechanical strength
  • High flow rate
  • Sterile, individually packed
  • All membrane filters have a high-contrast grid (3.1 mm)
  • Type STL in dispenser boxes for easy removal and secure handling with the Membrane-Butler
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
MicroPlus-21ST
Sterile
Black
Grid
Cellulose Nitrate
0.45 μm
Microbiological quality control
31mm
47 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers