Learn More
Sartorius Stedim Biotech™ Microsart™ Geneprep
Sartorius Stedim Biotech™ Microsart™ Geneprep is a reliable and rapid solution for DNA isolation and rehydration for contamination control purposes.
Brand: Sartorius Stedim Biotech™ SMB95-2004
This item is not returnable.
View return policy
Description
- Combines the steps of filtration of a liquid sample, dissolution of the membrane filter, lysis of the collected microorganisms, DNA isolation, and rehydration of the DNA in minimal volumes ready for PCR analysis, or any other DNA-based analysis method.
- Reliable, fast, and convenient.
- Unique membrane dissolving technology.
- Facilitates accurate detection of membrane-retained microbes.
Specifications
Microbial Detection | |
DNA-based detection | |
Material for 20 filtrations and 20 DNA extractions. | |
≥ 100 mL |
Product Content Correction
Product Title
Sartorius Stedim Biotech™ Microsart™ Geneprep > Reagents for 20 filtrations/20 extractions
Spot an opportunity for improvement?Share a Content Correction