missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Microsart™ Research Mycoplasma Real-Time PCR Kit
For fast and reliable screening of cell culture supernatants. Microsart™ Research Mycoplasma Real-Time PCR Kit is used for direct detection of Mollicutes in cell culture, media components, and derived biologicals.
Brand: Sartorius SMB95-1006
This item is not returnable.
View return policy
Description
- Enables fast and reliable detection of Mycoplasma DNA in cell culture supernatants
- Detects 16 S rRNA gene of at least 110 Mycoplasma species
- Time: Three hours
- Sample can be added directly to PCR reaction tube
- TaqMan™ real-time qPCR based chemistry
- Sample volume: 200μL
Applications:
- Used with any Real-time PCR instrument which detects the dyes FAM, ROX
- Used in quality control analysis in microbiological and biopharmaceutical labs
Compliance
EP 2.6.7
Specifications
2° to 8°C | |
100 tests | |
100 tests | |
Microbial Detection | |
Lyophilized |
qPCR | |
Assay test | |
16s rRNA of Mycoplasma | |
100 reactions | |
2μL |