Learn More
Carl Zeiss™ Microscope with Binocular Tube
Brand:  Carl Zeiss™ 491206-0006-000
Product Code. 10488624
  • £3900.00 / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Primo Vert available as predefined configurations, but additional configurations and accessories are available on request. A number of attractive features are designed to guarantee quick and reliable handling and ease of use:
  • Automatic light source switch-off (in addition to manual on/off-function): Primo Vert switches off the light automatically after 15min - saving energy and protecting the stand
  • Modular illumination: halogen or long lasting LED
  • High quality material: the stand is made of metal and all operating elements are coated with a special touch-friendly rubber compound
  • Universal phase slider (optional) for all objectives: for the ease of use there is one phase ring (Ph1) available for 10x, 20x and 40x - this means not having to change the position of the phase slider when changing objectives. In short: time-saving and economical
  • Increased working distance thanks to easy removal of the condenser, e.g., for roller-bottles
  • Objective indicators for fast identification of magnification
  • Integrated carrying handle
  • Award-winning design The new Primo Vert inverted microscope is focused on the essentials: excellent optics, high-durability quality materials, ease of use and an appealing design. The Primo Vert addresses a wide range of applications: from routine laboratories for live cell inspection to cutting edge research laboratories which require - in addition to research microscopes - compact and reliable routine microscopes for quick and efficient viewing of living cells. Typical applications include cancer and HIV research, human, animal and plant genetics, and cell biology in general.