missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Hazard Warning Tape
Repeats warning with hazard symbol for up to 66m. Azpack™ Hazard Warning Tape is self-adhesive and printed black on a yellow background.
£19.61 - £34.45
Specifications
Color | Black, Yellow |
---|---|
Format | Roll |
Material | Vinyl |
Quantity | 1 Ea. |
Length (Metric) | 66 m |
Product Code | Brand | Display Text | Dimensions (LxW) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Dimensions (LxW) | Price | Quantity & Availability | ||||
12318767
|
Azpack™
12318767 |
Danger Broken Glass | 66 m x 50 mm |
£19.61
Each |
In Stock
Log in to see stock. |
||||
10633405
|
Azpack™
10633405 |
Biohazard | 66 m x 25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Repeats warning with hazard symbol for up to 66m. Azpack™ Hazard Warning Tape is self-adhesive and printed black on a yellow background.
- Self-adhesive tape
- Wording and hazard symbol repeated along tape
- Black on yellow background
- 66m roll
- Cardboard core diameter: 75mm
- Widtch: 25mm
Specifications
Black, Yellow | |
Vinyl | |
66 m |
Roll | |
1 Ea. |