missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Microsep™ Advance Centrifugal Devices with Omega Membrane
Microsep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 5 mL of biological sample.
£184.00 - £680.00
Specifications
Description | EFA: 3.3 cm2 |
---|---|
Material (Membrane) | PES |
Sterility | Non-sterile |
Volume (Metric) | 0.5 to 5 mL (Sample) |
Product Code | Brand | MWCO | Includes | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Includes | Color | Price | Quantity & Availability | |||
17164741
|
Cytiva (formerly Pall Lab)
MCP001C41 |
1 kDa | 24/Pk. | Yellow |
£184.00
Pack of 24 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||
17134751
|
Cytiva (formerly Pall Lab)
MCP030C46 |
30 kDa | 100/Pk. | Red |
£680.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17194741
|
Cytiva (formerly Pall Lab)
MCP003C46 |
3 kDa | 100/Pk. | Gray |
£605.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17144751
|
Cytiva (formerly Pall Lab)
MCP100C41 |
100 kDa | 24/Pk. | Clear |
£232.00
Pack of 24 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||
17124751
|
Cytiva (formerly Pall Lab)
MCP030C41 |
30 kDa | 24/Pk. | Red |
£184.00
Pack of 24 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||
17154751
|
Cytiva (formerly Pall Lab)
MCP100C46 |
100 kDa | 100/Pk. | Clear |
£605.00
Pack of 100 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||
17114751
|
Cytiva (formerly Pall Lab)
MCP010C46 |
10 kDa | 100/Pk. | Blue |
£605.00
Pack of 100 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17184741
|
Cytiva (formerly Pall Lab)
MCP003C41 |
3 kDa | 24/Pk. | Gray |
£184.00
Pack of 24 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
17174741
|
Cytiva (formerly Pall Lab)
MCP001C46 |
1 kDa | 100/Pk. | Yellow |
£680.00
Pack of 100 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||
17104751
|
Cytiva (formerly Pall Lab)
MCP010C41 |
10 kDa | 24/Pk. | Blue |
£223.00
Pack of 24 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
Description
- Exchange buffer or remove salt of chromatography eluates and gradient fractions
- For research applications only
Specifications
EFA: 3.3 cm2 | |
PES | |
Non-sterile | |
0.5 to 5 mL (Sample) |