missing translation for 'onlineSavingsMsg'
Learn More

BD Microtainer™ Tubes with Microgard™ Closure

Product Code. p-8015497
Click to view available options
Closure Color:
Gold
Gray
Green
Lavender
Mint Green
Red
Includes:
Affected plain capillary tube
Affected plasma tube
Affected serum tube, UV prot.
Unaffected EDTA tube
Unaffected plasma tube
Unit Size:
Pack of 200
This item is not returnable. View return policy

Product Code. 12947646

Brand: BD 365966

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for whenever only the smallest amounts of blood are required. BD Microtainer™ Tubes with Microgard™ Closure are color coded to correspond with the type of blood collection and are marked with the correct filling level for the anticoagulant.

BD Microtainer™ tubes are used for the collection, transport and processing of capillary or venous blood from skin puncture. It is ideal for use with infants, children, geriatrics and emergency patients whenever only the smallest amounts of blood are required.
  • Wider diameter (8mm) for ease of collection and improved mixing
  • Color-coded caps correspond to the standard Vacutainer™ color-coding system for easy identification
  • Available with a variety of additives for use in various tests
  • Amber-tinted tubes are available for light sensitive analytes
  • Integrated collection scoop and visible fill lines
  • Microgard™ “twist assist” closure reduces the risk of splatter when removing the cap to protect against exposure
  • Closure can be used with a tube extender for use in 13mm x 75mm racks
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Product Type Microcollection Tube with Safety Closure
Additive Lithium Heparin
Material Polypropylene
Sterility Non-sterile
Closure Type Microgard
Capacity (Metric) 200 to 400 μL
Diameter (Metric) Inner 8 mm
Closure Included Yes
Closure Color Green
Quantity 200 Pk.
Includes Unaffected plasma tube
For Use With (Application) Collection, Transport and Processing of Skin Puncture (Capillary) Blood
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.