missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Microtiter Tube Plug Strip Product Code.: 12368202
Only undefined left
Add to basket

Thermo Scientific™ Microtiter Tube Plug Strip
GREENER_CHOICE

Product Code. 12368202 Shop All MBP Molecular BioProducts Products
Non-sterile, 120/Pk., 5/Cs., Pack of 120
Click to view available options
Sterility:
Non-sterile
Sterile
Quantity:
120/Pk., 5/Cs.
Unit Size:
Pack of 120
This item is not returnable. View return policy

Product Code. 12368202

Brand: Thermo Scientific™ 342611

Only undefined left
Add to basket
Only undefined left
Add to basket
This item is not returnable. View return policy

Seal 1.2mL microtiter tubes tightly for storage using these polyethylene plug strips.

Seal 1.2mL microtiter tubes tightly for storage by using Thermo Scientific™ Microtiter Tube Plug Strips.

  • LDPE non-autoclavable
  • Eight-plug strips can be easily cut apart for individual samples
  • Available sterile and nonsterile

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Product Type 8-cap Strip
Autoclavable Not Autoclavable
Description 1.2mL Microtiter Tube 8-cap plug strip
Material LDPE
Color Natural
Packaging 10 sealed bags of twelve 8 cap strips/unit, 5 units/case
Quantity 120/Pk., 5/Cs.
Sterility Non-sterile
For Use With (Equipment) Plug Strips
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Microtiter Tube Plug Strip >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.