missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Microtube Floating Storage Racks

Available in 24- or 48-place arrays with or without lids.

Brand:  Labcon™ 3035-500-000-9

 View more versions of this product

Product Code. 17537896

  • £22.10 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Holds 1.5, 1.7, or 2 mL microtubes
  • Freezable to -80°C
  • Made in USA
Specifications

Specifications

None
White
Polypropylene
Autoclavable
Microtubes (1.5, 1.7, 2 mL)
24-place
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers