Learn More
Corning™ Microvolume Pipet Tips, Nonsterile
Provide accurate, reliable performance in the stated volume range
Brand:  Corning™ 4901
 View more versions of this product
Product Code. 10387171
  • £399.65 / Pack of 10000
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.5 to 10 μL
Natural
Bulk
Bulk
Polypropylene
Non-sterile
DNase, RNase, and Pyrogen free
Microvolume, Round
Eppendorf Pipetter, Universal Fit Ultramicro