Learn More
Corning™ Microvolume Pipet Tips, Nonsterile
Provide accurate, reliable performance in the stated volume range
Brand: Corning™ 4826
This item is not returnable.
View return policy
Description
- Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
0.1 to 10 μL | |
Natural | |
Rack | |
Rack | |
Polypropylene |
Non-sterile | |
DNase, RNase, and Pyrogen free | |
Microvolume, Round | |
Gilson™ Pipetman™, Universal Fit Ultramicro |