Learn More
Trajan Scientific and Medical™ MicroVolume Syringes
Gives better performance and superior quality. Trajan Scientific and Medical MicroVolume Syringes has the knowledge and expertise that goes into manufacturing an SGE syringe.
Brand: Trajan Scientific and Medical™ 005055
This item is not returnable.
View return policy
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Manual injection syringe
Specifications
Syringe | |
0.20 mm | |
Removable Needle | |
70 mm |
25 G | |
0.5 mm | |
100 μL | |
Bevel |