Learn More
Kinesis™ Microwave Vials
Brand:  Kinesis™ VMW05-C-100-SB
Product Code. 15198102
  • £326.00 / Pack of 100
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
KX, for use with Initiator Systems (EXP only) from Biotage, contamination free 1st class hydrolytic borosilicate glass, resealable septa, 40°C to 300°C, 2.0mL to 5.0mL