missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Christeyns™ Mida Chriox 5
Mida Chriox 5 is a biodegradable, low residue broad spectrum sporicidal made of peracetic acid.
£148.00 - £808.00
Specifications
Product Type | Disinfectant Concentrate |
---|---|
Color-coded | No |
Form | Concentrated Liquid |
pH | Neutral |
Scent | Regular scent |
Product Code | Brand | Format | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Size | Price | Quantity & Availability | ||||
17831445
|
Christeyns™
695935 |
Can | 22 kg |
£148.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17841445
|
Christeyns™
695937 |
Drum | 222 kg |
£808.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Disinfectant based on peracetic acid
- Recommended for hygiene in cosmetic and pharmaceutical industries
- Suitable for the organic and cosmetic industries
- Usable in CIP without surfactants
- Measurement and control by conductivity
- Authorized for the following applications in: TP2: Disinfectant used in the private sector and in the field of public health and other biocidal products; TP4: Disinfectant for surfaces in contact with food and animal feed with mandatory rinsing
Specifications
Disinfectant Concentrate | |
Concentrated Liquid | |
Regular scent |
No | |
Neutral | |
Non-sterile |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Mida CHRIOX 5
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : hydrogen peroxide, acetic acid, peracetic acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Christeyns™ Mida Chriox 5