missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Midi Plus Horizontal Gel System

Includes 150mm x 70mm, 150mm x 100mm and 150mm x 150mm gel trays. Fisherbrand™ Midi Plus Horizontal Gel System can run up to 210 samples.

Brand:  Fisherbrand™ FMSCHOICETRIO

Product Code. 11833293

  • £501.28 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Supplied with 150 x 70mm, 150 x 100mm and 150 x 150mm gel trays
  • Run up to 210 samples
  • Low buffer volumes
  • Multichannel pipettor compatible combs for fast gel loading
  • Injection molded
  • Ideal for restriction fragment analysis, sample prep or checking of high numbers of samples.
Specifications

Specifications

Up to 210 Samples
150 x 70 mm, 150 x 100 mm and 150 x 150 mm gel trays
3
Horizontal Gel System
150mm x 70mm, 150mm x 100mm and 150mm x 150mm gel trays
2.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Midi Plus Horizontal Gel System > Length: 175mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.