missing translation for 'onlineSavingsMsg'
Learn More

Mifepristone, Tocris Bioscience™

Product Code. 30002128
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Progesterone and glucocorticoid receptor antagonist

Selective antagonist at progesterone (PR) and glucocorticoid (GR) receptors in vitro and in vivo. Is a silent antagonist at PR and has a higher affinity than progesterone. Has higher affinity for GR than dexamethasone (Cat. No. 1126).

Specifications

CAS 84371-65-3
Molecular Formula C29H35NO2
Quantity 100 mg
Synonym mifepristone, mifeprex, mifegyne, mifepriston, corlux, mifepristona, mifepristonum, korlym, mifepristonum latin, mifepristona spanish
InChI Key VKHAHZOOUSRJNA-GCNJZUOMSA-N
SMILES CC#CC1(CCC2C1(CC(C3=C4CCC(=O)C=C4CCC23)C5=CC=C(C=C5)N(C)C)C)O
IUPAC Name (8S,11R,13S,14S,17S)-11-[4-(dimethylamino)phenyl]-17-hydroxy-13-methyl-17-prop-1-ynyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 429.604
PubChem CID 55245
ChEBI CHEBI:50692
Formula Weight 429.6
Percent Purity >99%
Chemical Name or Material Mifepristone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'