missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MIIP/CEN1p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | migration and invasion inhibitory protein |
Gene Alias | IIP45 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073325
|
Abnova™
FG0204.100UL |
100 μL |
£1235.00
100µL |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083325
|
Abnova™
FG0204.200UL |
200 μL |
£2030.00
200µL |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
IIP45 | |
60672 | |
Liquid |
Tube | |
migration and invasion inhibitory protein | |
MIIP | |
DNA/RNA |