Learn More
Honeywell™ Millennia 2G Safety Glasses Product Code.: 15876665
In Stock
to see stock.
Add to basket
Honeywell™ Millennia 2G Safety Glasses
Product Code. 15876665
Clear
Click to view available options
Lens Tint:
Clear
Gray
Silver
Product Code. 15876665
Brand: Honeywell™ 1032179
In Stock
to see stock.
Add to basket
Add to basket
Safety glasses featuring excellent protection, comfortable fit, and stylish, simple design.
  • Softer, more flexible frame material for superior comfort against the head and ears when worn for all-day wear
  • Covers the eyes and areas above the cheeks and brow to ensure that the workers are protected from particulates, dust and other impact risks
  • A greater lens coverage wrap both peripherally and along the cheeks helps prevent particulate risk
  • A “shelf-like” design element along the brow has been added to help to prevent dust from dropping in between the frame and entering the eyes
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Description Dielectric
Certifications/Compliance (Europe) EC Category PPE: 2, EC Certificate: C1570.1HON, EC Attestation: C1570.1HON, EN C1570.1HON
Lens Tint Clear
For Use With (Application) For indoor use where eye protection is required against most commonly encountered hazards including impact protection and fogging.
Frame Color Black
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400
Product Content Correction
Product Title
Honeywell™ Millennia 2G Safety Glasses >