Learn More
Honeywell Safety Products™ Millenia Safety Glasses
Product Code. 12602633
Click to view available options
Frame Color:
Black
Blue
Silver
Lens Tint:
Clear
Silver Mirror
Yellow
Includes:
Adjustable, breakaway Flexicord retainer
Flexicord retainer
Lens Coating:
Impact-resistant, Anti-fog, UV-treated
Impact-resistant, Scratch-resistant, UV-treated
Unit Size:
Each
Product Code. 12602633
Brand: Honeywell Safety Products™ 1000009
Only null left
Add to basket
Add to basket
Virtually weightless eye protection for the active work environment. Honeywell Safety Products™ Millenia Safety Glasses block UV and provide protection for mechanical work including grinding, turning and milling.
  • General use and protection against impact (45m/s).
  • Suitable for mechanic work: grinding, turning, milling.
  • Polycarbonate lenses offer 99.9% UV protection up to 380nm.
  • Sensitive Tension Design and Parallel Vision Control.
  • A balanced fit and a greater visual comfort.
  • A virtually weightless feel.
  • Adjustable, breakaway Flexicord retainer.
  • Added security and convenience in the active work environment.
  • Unique 9 base curved lens design.
  • Excellent optical qualities and complete protection.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1131, EC Attestation Number EC1131
Lens Coating Impact-resistant, Scratch-resistant, UV-treated
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) Administration, Agriculture, Automotive, Building, Construction, Energy, Electricity, Laboratory, Logistics, Medical and Pharmaceutical and Wood industries
Frame Color Silver
Includes Adjustable, breakaway Flexicord retainer
No. per Pack 1 Ea.
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400