Learn More
Fisher BioReagents™ Miller's LB Agar, High Salt, Granulated
For propagation and maintenance of E. coli.
Brand: Fisher BioReagents™ 12821660
This item is not returnable.
View return policy
Description
- Specially granulated to reduce dust and increase solubility range
- Soluble in water
Specifications
Miller Luria-Bertani Broth | |
Agar | |
500 g |
Product Content Correction
Product Title
Fisher BioReagents™ Miller's LB Agar, High Salt, Granulated > Weight: 500g
Spot an opportunity for improvement?Share a Content Correction