Learn More
Fisher BioReagents™ LB Broth, High Salt, Granulated
For propagation and maintenance of E. coli.
Brand: Fisher BioReagents™ 12891650
Code : 32
Additional Details : Weight : 1.99800kg
Description
- Material specially granulated to reduce dust and increase solubility rate
- Sterilize by autoclaving
Specifications
Miller Luria-Bertani Broth | |
Microbiology Media | |
2000 g |