missing translation for 'onlineSavingsMsg'
Learn More

Merck Millex-GP Syringe Filter Unit

Millex-GP Syringe Filter Unit for sterilization of tissue culture media, protein solutions or aqueous solutions.

Brand:  Merck SE2M228I04

197.42 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.34000kg

Product Code. 10074574

  • £241.00 / Pack of 50
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

  • Sterile Millex filter units are available in a larger 33 mm housing with either MCE, PVDF or PES membranes.
  • Faster flow rate: The larger filter surface area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.
  • Higher operating pressure: Millex filters have a maximum housing pressure of 10.5 bar (150 psig), which means you can filter solutions faster than before.
  • Color-coded: The color-coded band on the Millex housing clearly indicates which membrane is inside.
  • Manufactured for reliable performance: Millex filters are manufactured in a controlled environment using an automated process. Human hands never touch the filter during assembly. A Certificate of Quality included in each box describes Millipore's quality standards in detail.
  • Choice of membranes: Millex filters are available with three membranes: Millipore Express™ (PES) membrane, mixed cellulose esters (MCE), and Durapore (PVDF).
  • Provides a unique combination of speed and six times less protein binding than competitive PES membranes. Millex-GP filters are recommended for sterile filtering protein solutions, tissue culture media, buffers and additives.
Specifications

Specifications

Green
PES, Modified Acrylic
Sterile
Low Protein Binding, Sterilization of drugs or aqueous solutions administered intravenously
Modified Acrylic
Male Luer Slip
45°C
200 mL
22 mm height
0.22 μm
4.5 cm2
< 0.1 mL
Female Luer-Lok
10 bar (145 psi)
Stem Cell Types: Mouse Embryonic Stem Cells, Human Embryonic Stem Cells, Mesenchymal Stem Cells, Neural Stem Cells, Hematopoietic Stem Cells, Epithelial Cells, Pancreatic Stem Cells, Cardiac Stem Cells, Induced Pluripotent Stem Cells
3.3 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers