Learn More
Merck Millicell μ-Angiogenesis Activation Assay Kit
Brand: Merck MMA130
This item is not returnable.
View return policy
Description
Specifications
Fluorescence | |
1 Kit | |
Angiogenesis Assays | |
Millicell μ-Angiogenesis Activation Assay Kit |
Millicell™ μ-Angiogenesis Slides Five tissue culture treated slides. Each slide contains 15 wells, Calcein-AM One 50 μg vial, PMA One 1mg vial. ITS Media Supplement, 100X One 0.5mL vial, Thrombin Solution One 0.75mL vial, Fibrinogen Solution | |
1 Kit | |
Vertebrates, Human, Rat |