Learn More
Mineral Oil Rotational Viscosity Standard, Paragon Scientific™
Paragon ISO 17025 / ISO Guide 34 certified mineral oil rotational viscosity standards are specifically formulated for use with rotational viscometers.
Brand: Paragon Scientific RTM13
This item is not returnable.
View return policy
Description
- Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in strict accordance with ASTM D2162 at 20°C and 25°C
- Density g/mL in accordance with ASTM D1480 at 20°C and 25°C
- Kinematic viscosity at other temperatures are calculated in accordance with ASTM D341
- Densities at other temperatures are derived from proportional calculations of the measurements at 20°C and 25°C
- 500mL inert glass packaging with tamper evident cap provides absolute assurance of sample integrity
- Full traceability to National Standards
- 12 month shelf life
- Manufactured in the United Kingdom
Specifications
500 mL | |
75.19 mPa.s (25°C) | |
Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation |
Product Content Correction
Product Title
Nominal values quoted
Spot an opportunity for improvement?Share a Content Correction