Learn More
Thermo Scientific™ Mini Capillary Unions
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Brand:  Thermo Scientific™ 290GU499
 View more versions of this product
Product Code. 10289074
  • £57.69 / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Kit contains one union and five ferrules
Includes:
Union, 1; Ferrules, 5
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Capillary Union
For 0.53 mm I.D. Capillary
0.53 mm ID Capillary GC Column