Learn More
Neogen Mini Flip-Top Vial
Product Code. 17298520
Encompass
Click to view available options
Includes:
9 mL Letheen Broth
9 mL Maximum Recovery Diluent
Buffered Peptone Water Broth
Unit Size:
Pack of 100
Product Code. 17298520
Brand: Neogen 7000030317
Leak-resistant plastic cylinder with a flip-open lid that seals securely for safe shipping and handling.
  • Hinged lid can be opened with one hand and stays open for easy sample additions.
  • Decontaminated through gamma irradiation and ready for use with no additional blending required.
  • Flat bottom tube stands on its own, no holder required.
  • Leak resistant vial helps prevent leakage during shipping.
  • 2 year shelf life from date of manufacture provides long-term readiness.
Specifications
Test Type Food and Beverage, Environmental Test
Includes 9 mL Maximum Recovery Diluent
Quantity 100/Cs.