Learn More
Pyrex™ Mini Gas Bubbler
Used in inert atmosphere experiments. Pyrex™ Mini Gas Bubbler is ideal for bubble counting. Includes detachable plastic hose connections to allow easy and safe attachment to 9mm bore flexible tubing.
Brand: Quickfit GB3/SC
Code : Z4
Additional Details : Weight : 0.04000kg
Description
- For use in inert atmosphere experiments
- Ideal for bubble counting
- Complete with detachable plastic hose connections to allow easy and safe attachment to 9mm bore flexible tubing