Learn More
Invitrogen™ Mini Gel Tank and Blot Module Set
Provides all of the necessary devices for protein separation and Western blotting using Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels
Brand: Invitrogen™ NW2000
669.80 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes: Mini Gel Tank (Runner Tank; Cassette Clamps, left and right; Tank Base; Tank Lid; and Power Supply Adapters) and two Mini Blot Modules (each with Mini Blot Module with Gasket, Four Blotting Sponges, Blotting Tweezer, Blotting Roller)
Description
This set contains a Mini Gel Tank for protein separation and two Mini Blot Modules for Western transfer.
Mini Gel Tank Features
- Compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt Bis-Tris Plus gels
- Accommodates up to two gels per run
- Tank design enables side-by-side gel loading and enhanced viewing during use
- Optimized conditions using constant voltage allow Bolt Bis-Tris Plus gels to be run in approximately 35 min.
- Run times vary depending on buffer conditions and power supply being used
- Also accommodates one blot module per chamber, or two blot modules total with side-by-side layout, for convenient Western transfers
Mini Blot Module Features
- Wet transfer device used exclusively with the Mini Gel Tank
- Compatible with Bolt Bis-Tris Plus Gels, accomodating one gel per transfer
- Universal connection and molded gasket make the blot module easy to use
- Inner core of blot module allows for use of less methanol-based transfer buffer per transfer
- At recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes typically in 30 to 60 min.
The current model Mini Gel Tank includes cassette clamps with gray cam handles designed for use with the 10cm Bolt minigels as well as NuPAGE and Novex Tris-Glycine minigels.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
• 1 mini gel tank • 2 blot modules |
|
Novex™ Mini Gels | |
Invitrogen | |
Mini Gel Tank and Blot Module Set | |
Room Temperature |
2 Gels/Run | |
Mini | |
35 min. | |
1 set | |
500 VDC |
Special Offers
-
Multiple Manufacturers
New Offer Shop Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.