missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Mini Pictograms Mix
Safety icons designed to improve workplace safety.
Brand: Brady™ 814013
11.96 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- 1 sheet contains 11 off the following icons:
- PIC1801: Explosive
- PIC1803: Oxodizing
- PIC1804: Compressed Gas
- PIC1805: Health Hazard
- PIC1806: Hazardous to Aquatic Enviroment
- PIC1807: Respiratory Hazard
- PIC1808: Corrosive
- PIC1809: Acute Toxicity
- And 16 of:
- PIC1802: Flammable
Specifications
Gloss | |
Permanent Acrylic | |
10 x 10 mm |
Magnetic Plastic | |
Diamond | |
Black |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Mini Pictograms Mix > Dimensions (L x W): 10 x 10 mm
Spot an opportunity for improvement?Share a Content Correction