Learn More
IBA Lifesciences Mini Spin Columns
For rapid purification of recombinant proteins
£24.65 - £95.35
Specifications
For Use With (Application) Rapid Purification of Recombinant Proteins
View More Specs
Products 2
Product Code Brand Kit Contents Price Quantity & Availability  
Product Code Brand Kit Contents Price Quantity & Availability  
17257590
View Documents Promotion Details
IBA Lifesciences
2-9102-010
10 columns This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17267590
View Documents Promotion Details
IBA Lifesciences
2-9102-050
50 columns This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Mini Spin Columns are the ideal solution to treat small volumes in combination with Selector Resins.
  • They fit into every microcentrifuge and are compatible with all standard 1.5 ml or 2.0 ml tubes.
  • Mini Spin Columns are provided with inserted 10 μm filters, screw caps, and snap-off plugs
Specifications
Specifications
Rapid Purification of Recombinant Proteins