missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Mini Tube System

Polypropylene tube system for compound library storage

Brand:  Axygen™ MTS-11-8-C-R-S

411.13 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12716829

  • £449.00 / Pack of 600
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.

  • Rack features a one-way lid design and alpha-numeric grid
  • Molded-in condensation rings on the lid minimize cross contamination
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Clear
Ensures positive sample identification
120/Pk.
Sterile
Cylindrical
1.1 mL
Polypropylene
Alpha numeric grid, one way lid
12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs.
No
Microtube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.