missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ V4A Stainless Steel Mini ViscoSampler

Buerkle™ V4A Stainless Steel Mini ViscoSampler for viscous substances such as pastes, mud, creams, oils.

Brand:  Buerkle™ 5341-1150

416.71 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10421731

  • £454.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel AISI 316L (1.4404), seals PTFE
  • Sterilisable
  • Ideal for the pharmaceutical and cosmetics industry
  • The sample is drawn in using the pump rod and directly conveyed to the sampling container
  • Easy cleaning for flawless sampling
  • Depending on the viscosity, the removable suction tip can be fitted with suction drills of 2mm to 6mm
Specifications

Specifications

Mini Viscosampler
Autoclavable
2.54 oz.
Ideal for The Pharmaceutical and Cosmetics Industry
1500 mm
Stainless Steel (V4A), PTFE
2 mm Suction tip
75 mL
1500 x 15 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers