Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™
Versatile check valves can be installed in any position
£3.10 - £4.10
Specifications
Outlet Port Barbed
Material (Seal) Viton
View More Specs
Products 4
Product Code Brand Inlet Price Quantity & Availability  
Product Code Brand Inlet Price Quantity & Availability  
30054373
View Documents
Cole-Parmer
50115-48
1/4 in. hose barb
£4.10
Each
Estimated Shipment: 17-10-2025
to see stock.
Add to basket
30054130
View Documents
Cole-Parmer
50115-20
1/8 in. hose barb
£3.10
Each
Estimated Shipment: 17-10-2025
to see stock.
Add to basket
30053078
View Documents
Cole-Parmer
50115-21
3/16 in. hose barb
£4.10
Each
Estimated Shipment: 17-10-2025
to see stock.
Add to basket
30054160
View Documents
Cole-Parmer
50115-22
5/16 in. hose barb
£4.10
Each
Estimated Shipment: 17-10-2025
to see stock.
Add to basket
Description
Description
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. 0.18 psi breaking pressure
Specifications
Specifications
Barbed
Viton
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™