Learn More
Greiner Bio-One MiniCollect™ Tube. Lavender Cap, K2E K2EDTA, Non-Sterile
Inner wall is coated with anticoagulant that binds calcium ions and interrupts clotting.
Brand: Greiner Bio-One 450532
Description
- Inner wall is coated with spray-dried K2E K2EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.

Specifications
K2 E K2 EDTA | |
0.25, 0.5 mL | |
Lavender |
Non-sterile | |
Yes | |
500/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- X500 MiniCollect TUBE 0.25 / 0.5 ml K2E K2EDTA la vender cap
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Ethylenediaminetetraacetic Acid Tripotassium Dihydrate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.