missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One MiniCollect™ Tube. Lavender Cap, K3E K3EDTA, Non-Sterile

Product Code. 15830146
Click to view available options
Capacity (Metric):
0.25, 0.5 mL
1 mL
Unit Size:
Pack of 2000
Pack of 500
This item is not returnable. View return policy

Product Code. 15830146

Brand: Greiner Bio-One 450530

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inner wall is coated with anticoagulant that binds calcium ions and interrupts clotting.

  • Inner wall is coated with spray-dried K3E K3EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.
  • Used for hematology.
  • Available in different quantities.
TRUSTED_SUSTAINABILITY

Specifications

Additive K3 E K3 EDTA
Sterility Non-sterile
Capacity (Metric) 0.25, 0.5 mL
Closure Included Yes
Closure Color Lavender
compliance-icons
Product Identifier
  • X500 MiniCollect TUBE 0.25/0.5 ml K3E K3EDTA lave nder cap
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ethylenediaminetetraacetic Acid Tripotassium Dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.