missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One MiniCollect™ Tube, Light-Blue Cap, 9NC Coagulation Sodium Citrate 3.2%, Sterile
Contains a buffered sodium citrate solution to aid in examination of coagulation parameters.
£1133.47
Specifications
Additive | 9NC Sodium Citrate 3.2% |
---|---|
Sterility | Sterile |
Closure Included | Yes |
Closure Color | Light Blue |
Quantity | 500/Pk. |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15738179
|
Greiner Bio-One
450539 |
1 mL |
£1133.47
Pack of 500 |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Contains a buffered sodium citrate solution to aid in examination of coagulation parameters.
- This light blue top sodium citrate tube, or coagulation tube, is used for examination of coagulation parameters.
- Coagulation tubes contain a buffered sodium citrate solution.
Specifications
9NC Sodium Citrate 3.2% | |
Yes | |
500/Pk. |
Sterile | |
Light Blue |