missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Packaging™ Minigrip™ Polyethylene Resealable Bags
Manufactured from polyethylene. Alpha Packaging™ Minigrip™ Polyethylene Resealable Bags are ideal for sending samples.
£52.84 - £74.06
Specifications
For Use With (Application) | Ideal for storing stationery and sending samples |
---|---|
Material | Polyethylene |
Thickness (Metric) | 50 μm |
Quantity | 100 Pack |
Product Code | Brand | Width (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
12380529
|
Alpha Packaging™
12380529 |
150 mm | 140 mm |
£74.06
Pack of 100 |
In Stock
Log in to see stock. |
||||
12390549
|
Alpha Packaging™
12390549 |
380 mm | 510 mm |
£52.84
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- Resealable section for the specimen
- Open pocket for the associated documents.
- Plain bag; no biohazard warning label
- Opening along width
- 50μm thickness.
- Polyethylene
- Three write-on bars for labelling
Specifications
Ideal for storing stationery and sending samples | |
Polyethylene | |
50 μm | |
100 Pack |