missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ MiniMark™ Vinyl Indoor Tape
MiniMark™ Vinyl Indoor Tape suitable for logistics identification.
£117.00 - £147.00
Specifications
Description | MiniMark Indoor Vinyl Tape |
---|---|
Adhesive Type | Permanent Acrylic, Permanent |
Color | White |
Finish | Gloss |
Format | Roll |
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
11540014
|
Brady™
710065 |
35 m x 50 mm |
£147.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||||
11530014
|
Brady™
710063 |
35 m x 25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Indoor vinyl tape suitable for logistics identification.
Specifications
MiniMark Indoor Vinyl Tape | |
White | |
Roll | |
Thermal Transfer | |
Facility Identification, Floor marking, Hazard communication Identification, Pipe and valve marking, Safety Identification, Warehouse marking | |
1 | |
-40°C | |
1 Ea. | |
-40°C to +70°C | |
0.102 mm | |
35 m |
Permanent Acrylic, Permanent | |
Gloss | |
Vinyl | |
Labels | |
MiniMark Printer | |
2 Years | |
1 Roll/Box | |
-40°F to +158°F | |
0.004 in. | |
115 ft. | |
1.1 kg |