Learn More
Cytiva (formerly Pall Lab) Minimate™ Tangential Flow Filtration (TFF) Capsule with Omega™ Membrane
Concentration and diafiltration (desalting or buffer exchange) processes can be performed on the same system with minimal user intervention.
Brand:  Cytiva (formerly Pall Lab) OA300C12
Additional Details : Weight : 0.46550kg
 View more versions of this product
Product Code. 17144771
  • £324.00 / Each
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
  • Efficient processing
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
PES
Non-sterile
1/Pk.
300 kDa
50 cm2