Learn More
Gilson™ Speed Control Module
Integrates easily into existing systems with direct control through the keypad or remotely through another device. The Speed Control Module is quiet and user friendly. For use with MINIPULS™ 3 Peristaltic Pump.
Brand:  Gilson™ F155001
1971.09 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 10295492
  • £2380.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quiet, user friendly and easy to integrate into your existing system with direct control through the keypad or remotely through another device.
  • For use with MINIPULS™ 3 Peristaltic Pump.
  • Flow Rate: 0.3 μL to 220 mL/min.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MINIPULS™ 3
High-torque Stepper Motor
Microprocessor
Module
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ Speed Control Module > Speed Control Module; Flow Rate: 0.3 μL to 220 mL/min.