Learn More
Cytiva™ MiniTrap™ Column
The Cytiva™ MiniTrap™ Columns are small-scale chromatography columns designed for rapid and convenient purification. They are commonly used for various applications including protein purification, antibody purification, peptide purification and nucleic acid purification.
Brand: Cytiva™ 28-9409-28
Code : GX
Additional Details : Weight : 0.53900kg
Description
- For rapid and efficient separation and purification of samples
- Single use, pre-packed, disposable format allows for easy and convenient sample processing
- Ideal for use in R and D, process development and QC laboratories
Specifications
MiniTrap chromatography column | |
Small-scale purification and separation | |
Single-use |
Column | |
Chromatography Cartridge | |
1 Ea. |
Safety and Handling
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-2H-isothiazol-3-one