missing translation for 'onlineSavingsMsg'
Learn More

Minocycline hydrochloride, MedChemExpress

Product Code. p-200102037
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16433347

Brand: Medchem Express HY17412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Minocycline hydrochloride is an orally active, potent and BBB-penetrated semi-synthetic tetracycline antibiotic.

Minocycline hydrochloride is a hypoxia-inducible factor (HIF)-1α inhibitor. Minocycline hydrochloride shows anti-cancer, anti-inflammatory, and glutamate antagonist effects. Minocycline hydrochloride reduces glutamate neurotransmission and shows neuroprotective properties and antidepressant effects. Minocycline hydrochloride inhibits bacterial protein synthesis through binding with the 30S subunit of the bacterial ribosome, resulting in a bacteriostatic effect.

Specifications

Chemical Name or Material Minocycline hydrochloride
CAS 13614-98-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H28ClN3O7
Quantity 50 mg
Solubility Information H2O : 10 mg/mL (20.25 mM; Need ultrasonic) ∣DMSO : 0.49 mg/mL (0.99 mM; Need ultrasonic and warming)
SMILES OC1=C(C(N)=O)C([C@@]2(O)C(O)=C3C(C4=C(O)C=CC(N(C)C)=C4C[C@@]3([H])C[C@@]2([H])[C@@H]1N(C)C)=O)=O.Cl
Molecular Weight (g/mol) 493.94
Formula Weight 493.94
Percent Purity 98.47%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.