missing translation for 'onlineSavingsMsg'
Learn More

Minoxidil, MedChemExpress

Product Code. 16440778
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16440778

Brand: Medchem Express HYB0112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Minoxidil (U10858) is an ATP-sensitive potassium (KATP) channel opener, a potent oral antihypertensive agent and a peripheral vasodilator that promotes vasodilation also affects hair growth. Minoxidil is also a potent inhibitor of soybean lipoxygenaseare with an IC50 of 20 μM.

Minoxidil (U10858) is an ATP-sensitive potassium (KATP) channel opener, a potent oral antihypertensive agent and a peripheral vasodilator that promotes vasodilation also affects hair growth. Minoxidil is also a potent inhibitor of soybean lipoxygenaseare with an IC50 of 20 μM.

Specifications

Chemical Name or Material Minoxidil
Physical Form Powder
CAS 38304-91-5
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C9H15N5O
For Use With (Application) COVID-19-immunoregulation
Synonym U10858
Solubility Information Ethanol : 7.14 mg/mL (34.12 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.78 mM; ultrasonic and warming and heat to 50°C)
SMILES NC1=NC(N2CCCCC2)=CC(N)=[N+]1[O-]
Molecular Weight (g/mol) 209.25
Formula Weight 209.25
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.