missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mirin, 95%

Product Code. 15488739
Click to view available options
Quantity:
5 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299953-00-7
Molecular Formula C10H8N2O2S
Synonym cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone;

Specifications

Quantity 10 mg
Description Orange
Content And Storage -30°C to -10°C
Formula Weight 220.25g/mol
Sensitivity Moisture Sensitive
Form Solid
InChI Key YBHQCJILTOVLHD-UHFFFAOYSA-N
SMILES NC1=NC(=O)C(S1)=CC1=CC=C(O)C=C1
IUPAC Name 2-amino-5-[(4-hydroxyphenyl)methylidene]-4,5-dihydro-1,3-thiazol-4-one
Molecular Weight (g/mol) 220.25
Assay 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'